Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
ATP2A2 Rabbit anti-Human, Unconjugated, Clone: AOGE-1, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB20700
Catalog No. 89-337-100
Description
Description
ATP2A2 Monoclonal antibody specifically detects ATP2A2 in Human samples. It is validated for ICC, IHC, WB-Ce, IF, Flow Cyt.
Specifications
Specifications
ATP2A2
AOGE-1
ATPase, Ca++ transporting, cardiac muscle, slow twitch 2
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
ATP2A2
ATP2B, DAR, DD, DKFZp686P0211, FLJ20293, FLJ38063, MGC45367, SERCA2
A synthetic peptide corresponding to human ATP2A2
100 μg
Primary
488