Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5B Mouse anti-Canine, Human, Rat, Clone: OTI4E5, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500833

Catalog No. 50-167-1979


Add to cart

Description

Description

ATP synthase is extremely conserved through evolution and can be found in plants, fungi, bacteria, and animals. The ATP synthase enzyme is a transmembrane protein responsible for driving the reversible reaction from ADP+ phosphate to ATP. This reaction is accomplished by a flux of protons across the membrane as a result of electron transfer. The ATP synthase protein has two main sections; the F1 ATP-ase (soluble) and the F0 ATP-ase (membrane embedded). The F1 section consists of the alpha, beta, gamma, delta, and epsilon subunits. While the F0 consists of a, b, and c subunits.
Specifications

Specifications

ATP5B
Monoclonal
0.9 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P06576, P10719, P99504
ATP5B, ATPMB, ATPSB, HEL-S-271
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
171374, 403669, 506
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI4E5
Unconjugated
ATP5B
Liquid
ATP5B
Full length human recombit protein of human ATP5B produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit