Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

ATP5I Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Antigen ATP5I
Dilution Western Blot 1:500-1:2000, Immunohistochemistry -1:500-1:2000, Immunoprecipitation -1-5-ul/mg, Immunohistochemistry-Paraffin -1:500-1:2000
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121477
View Documents
Novus Biologicals
NBP31138450UL
50 μg
Each for $138.37
N/A
NB121476
View Documents
Novus Biologicals
NBP311384200UL
200 μg
Each for $320.71
N/A
Description

Description

ATP5I Polyclonal antibody specifically detects ATP5I in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

ATP5I
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Purified
RUO
PBS, pH7.0
521
IgG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000, Immunohistochemistry -1:500-1:2000, Immunoprecipitation -1-5-ul/mg, Immunohistochemistry-Paraffin -1:500-1:2000
Unconjugated
Rabbit
Human
ATP synthase e chain, mitochondrial, ATP synthase subunit e, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit E, ATP synthase, H+ transporting, mitochondrial Fo complex, subunit E, ATP5K, ATPase subunit e, F1F0-ATP synthase, murine e subunit, MGC12532
E. coli-derived Human ATP5I fragment
Polyclonal
Antigen and protein A Affinity-purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit