Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5O Antibody (OTI2E9), DyLight 755, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270239IR

Catalog No. NBP270239IR


Add to Cart

Description

Description

ATP5O Monoclonal specifically detects ATP5O in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ATP5O
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ATP5PO
Recombinant protein fragment corresponding to amino acids 24-213 of human ATP5O (NP_001688) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI2E9
Western Blot, Immunohistochemistry
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit, human ATP synthase OSCP subunit, oligomycin sensitivity conferring protein10ATPOATP synthase subunit O, mitochondrial, Oligomycin sensitivity conferral protein, oligomycin sensitivity conferring protein, OSCPhuman ATP synthase OSCP subunit
Mouse
Affinity Purified
Core ESC Like Genes, Stem Cell Markers
539.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit