Learn More
Abnova™ ATP6V0B 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000533T04
Catalog No. 89-936-055
Add to cart
Description
Description
Specifications
Specifications
293T
NM_004047.3
pCMV-ATP6V0B full-length
100 μL
Human
ATP6V0B 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 21.4kDa
Transient overexpression cell lysate was tested with anti-ATP6V0B antibody (Mfr. No. H00000533-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot