Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V0E2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | ATP6V0E2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155120UL
|
Novus Biologicals
NBP15510020UL |
20 μL |
Each for $152.22
|
|
NBP155100
|
Novus Biologicals
NBP155100 |
100 μL |
Each for $436.00
|
|
Description
ATP6V0E2 Polyclonal specifically detects ATP6V0E2 in Human samples. It is validated for Western Blot.Specifications
ATP6V0E2 | |
Polyclonal | |
Rabbit | |
Human | |
155066 | |
Synthetic peptides corresponding to ATP6V0E2(ATPase, H+ transporting V0 subunit e2) The peptide sequence was selected from the middle region of ATP6V0E2. Peptide sequence TVAPLSLTTPSSGPSPTQLCLVTSSLLLAPRDPDPQGLPGSWKSSQSSQP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ATP6V0E2 | |
IgG | |
Affinity Purified | |
17-19 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title