Learn More
ATP6V1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150971AP
Catalog No. 50-172-9975
Add to cart
Description
Description
ATP6V1B2 is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A, three B, and two G subunits, as well as a C, D, E, F, and H subunit. The V1 domain contains the ATP catalytic site. ATP6V1B2 is one of two V1 domain B subunit isoforms and is the only B isoform highly expressed in osteoclasts.
Specifications
Specifications
ATP6V1B2
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21281, P62814, P62815
ATP6B1B2, ATP6B2, ATP6V1B2, HO57, V ATPase subunit B 2, VATB, Vma2, VPP3
Rabbit
IgG
150 μL
-20°C
Primary
117596, 11966, 526
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ATP6V1B2
Liquid
ATP6V1B2
ATP6V1B2 Fusion Protein Ag7174
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat