Learn More
ATP6V1C1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160541AP
Catalog No. 50-173-0680
Add to cart
Description
Description
ATP6V1C1 is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c ft., c ft. ft., and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene is one of two genes that encode the V1 domain C subunit proteins and is found ubiquitously. This C subunit is analogous but not homologous to gamma subunit of F-ATPases. Previously, this gene was designated ATP6D.
Specifications
Specifications
ATP6V1C1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21283, Q5FVI6, Q9Z1G3
ATP6C, ATP6D, ATP6V1C1, V ATPase subunit C 1, VATC, Vma5
Rabbit
IgG
150 μL
-20°C
Primary
299971, 528, 66335
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ATP6V1C1
Liquid
ATP6V1C1
ATP6V1C1 Fusion Protein Ag9020
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat