Learn More
ATP6V1H Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 266831AP
Catalog No. 50-114-2760
Add to cart
Description
Description
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular organelles. V-ATPase-dependent organelle acidification is necessary for multiple processes including protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. The encoded protein is the regulatory H subunit of the V1 domain of V-ATPase, which is required for catalysis of ATP but not the assembly of V-ATPase. Decreased expression of this gene may play a role in the development of type 2 diabetes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
Specifications
ATP6V1H
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UI12
ATP6V1H, CGI 11, MSTP042, NBP1, Nef binding protein 1, SFD, SFDalpha, SFDbeta, V ATPase 50/57 kDa subunits, V ATPase subunit H, V type proton ATPase subunit H, Vacuolar proton pump subunit H, VMA13
Rabbit
IgG
150 μL
-20°C
Primary
51606
Immunoprecipitation, Western Blot
Unconjugated
ATP6V1H
Liquid
ATP6V1H
ATP6V1H Fusion Protein Ag24688
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human