Learn More
ATP7A Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: S60-4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259376UV
Catalog No. NBP259376UV
Add to cart
Description
Description
ATP7A Monoclonal antibody specifically detects ATP7A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ATP7A
S60-4
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
ATP7A
Synthetic peptide amino acids 42-61 (cytoplasmic C-terminus) of human Copper- transporting ATPase1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects the extracellular domain of LRP4 215kDa. Degradation product can be seen at approx 150kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
ATPase, Cu++ transporting, alpha polypeptide, Copper pump 1, copper-transporting ATPase 1, Cu++-transporting P-type ATPase, DSMAX, EC 3.6.3, EC 3.6.3.4, MC1, Menkes disease-associated protein, Menkes syndrome, MK, MNKFLJ17790, OHS, SMAX3
Mouse
IgG2b
100 ul
Primary
538
Human, Mouse, Rat