Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP7B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA116584
Description
This antibody does not react with mouse samples.
This gene is a member of the P-type cation transport ATPase family and encodes a protein with several membrane-spanning domains, an ATPase consensus sequence, a hinge domain, a phosphorylation site, and at least 2 putative copper-binding sites. This protein functions as a monomer, exporting copper out of the cells, such as the efflux of hepatic copper into the bile. Alternate transcriptional splice variants, encoding different isoforms with distinct cellular localizations, have been characterized. Mutations in this gene have been associated with Wilson disease (WD).
Specifications
ATP7B | |
Polyclonal | |
Unconjugated | |
Atp7b | |
Atp7a; ATP7B; ATPase copper transporting beta; ATPase, Cu(2+)- transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide (same as Wilson disease); copper pump 2; Copper-transporting ATPase 2; Hts; PINA; PINA gene, promoter; pineal night-specific ATPase; PWD; RP11-327P2.1; RP11-327P2.3; toxic milk; tx; WC1; WD; Wilson disease-associated protein; wilson disease-associated protein homolog; Wilson protein; Wnd; WND/140 kDa | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24218, 540 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P35670, Q64535 | |
Atp7b | |
Synthetic peptide corresponding to an internal sequence near the C-terminus of human ATP7b. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction