Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATP8A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114355
Description
ATP8A1 Polyclonal Antibody for Western Blot, Flow
The P-type adenosinetriphosphatases (P-type ATPases) are a family of proteins which use the free energy of ATP hydrolysis to drive uphill transport of ions across membranes. Several subfamilies of P-type ATPases have been identified. One subfamily catalyzes transport of heavy metal ions. Another subfamily transports non-heavy metal ions (NMHI). The protein encoded by this gene is a member of the third subfamily of P-type ATPases and acts to transport amphipaths, such as phosphatidylserine. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
ATP8A1 | |
Polyclonal | |
Unconjugated | |
ATP8A1 | |
AI481521; AI853962; aminophospholipid translocase; APLT; Atp3a2; Atp8a1; ATPase 8A1, aminophospholipid transporter (APLT), class I; ATPase 8A1, p type; ATPase class I type 8A member 1; ATPase II; ATPase phospholipid transporting 8A1; ATPase, aminophospholipid transporter (APLT), class I, type 8A, member 1; ATPASEII; Atpc1; ATPIA; ATPP2; AW743152; AW822227; B230107D19Rik; Chromaffin granule ATPase II; ClassI; P4-ATPase flippase complex alpha subunit ATP8A1; phospholipid-transporting ATPase IA; probable phospholipid-transporting ATPase IA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10396, 289615 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y2Q0 | |
ATP8A1 | |
Recombinant protein within Human ATP8A1 aa 410-637. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction