Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP8B3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120588

This item is not returnable.
View return policy
Description
Positive test controls include: HCT116.
ATP8B3 Polyclonal antibody specifically detects ATP8B3 in Human samples. It is validated for Western BlotSpecifications
ATP8B3 | |
Polyclonal | |
Unconjugated | |
ATP8B3 | |
aminophospholipid translocase ATP8B3; ATP1K; ATP8B3; ATPase class I type 8B member 3; ATPase phospholipid transporting 8B3; ATPase, aminophospholipid transporter, class I, type 8B, member 3; ATPase, class I, type 8B, member 3; ATPIK; FOS37502_2; Phospholipid-transporting ATPase IK; potential phospholipid-transporting ATPase IK; probable phospholipid-transporting ATPase IK | |
Recombinant fusion protein containing a sequence corresponding to amino acids 600-900 of human ATP8B3 (NP_620168.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
2.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60423 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
148229 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title