Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPA, Tocris Bioscience™

Catalog Number p-4783269
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
140158-50-5
C10H16N2O4
228.248
PIXJURSCCVBKRF-UHFFFAOYSA-N
atpa, atpa-tert, r,s-atpa, alpha-amino-3-hydroxy-5-tert-butyl-4-isoxazolepropionate, 2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propionic acid, rs-2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo-4-isoxazolepropanoic acid, 4-isoxazolepropanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo, atpa, solid, acmc-20cirp
2253
2-amino-3-(5-tert-butyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
CC(C)(C)C1=C(C(=O)NO1)CC(C(=O)O)N
This item is not returnable. View return policy
140158-50-5
C10H16N2O4
228.248
PIXJURSCCVBKRF-UHFFFAOYSA-N
atpa, atpa-tert, r,s-atpa, alpha-amino-3-hydroxy-5-tert-butyl-4-isoxazolepropionate, 2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propionic acid, rs-2-amino-3-3-hydroxy-5-tert-butylisoxazol-4-yl propanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo-4-isoxazolepropanoic acid, 4-isoxazolepropanoic acid, alpha-amino-5-1,1-dimethylethyl-2,3-dihydro-3-oxo, atpa, solid, acmc-20cirp
2253
2-amino-3-(5-tert-butyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
CC(C)(C)C1=C(C(=O)NO1)CC(C(=O)O)N

Potent and selective GluK1 kainate agonist

A selective and potent GluK1 (formerly GluR5) kainate receptor agonist (Ki = 4.3 nM), inactive at GluK6 (formerly GluR6) (Ki > 1 mM) and only weakly active at AMPA receptors (GluA1-4) and the kainate receptors GluK5 (formerly KA-2) and GluK3 (formerly GluR7) (Ki values of 6 - 14 μM).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Quantity 10 mg
Formula Weight 228.25
Percent Purity >98%
Chemical Name or Material ATPA
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit