Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPase Na+/K+ beta 2 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$177.29 - $436.00

Specifications

Antigen ATPase Na+/K+ beta 2
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094866
View Documents
Novus Biologicals
NBP303790100UL
100 μL
Each for $436.00
Add to cart
 
NB094867
View Documents
Novus Biologicals
NBP30379020UL
20 μL
Each for $177.29
Add to cart
 
Description

Description

ATPase Na+/K+ beta 2 Polyclonal specifically detects ATPase Na+/K+ beta 2 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ATPase Na+/K+ beta 2
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
482
IgG
Affinity purified
Immunohistochemistry
Unconjugated
Rabbit
Mouse, Rat
adhesion molecule on glia, AMOGsodium/potassium-dependent ATPase beta-2 subunit, ATPase, Na+/K+ transporting, beta 2 polypeptide, Na, K-ATPase beta-2 polypeptide, Sodium/potassium-dependent ATPase subunit beta-2, sodium/potassium-transporting ATPase beta-2 chain, sodium/potassium-transporting ATPase subunit beta-2
Recombinant fusion protein containing a sequence corresponding to amino acids 68-290 of human ATPase Na+/K+ beta 2 (NP_001669.3).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit