Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPB Mouse anti-Human, Rat, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI4E5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270224APCCY7

Catalog No. N270224APC7


Add to cart

Description

Description

ATPB Monoclonal antibody specifically detects ATPB in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

ATPB
OTI4E5
Flow Cytometry
Affinity Purified, Conjugated
ATP5F1B
Full length recombinant protein of human ATPB (NP_001677) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Rat
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit
Mouse
IgG2a
0.1 mL
Primary
506.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit