Learn More
Abnova™ ATPBD1B 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00054707T01
Catalog No. 89-936-130
Add to cart
Description
Description
Specifications
Specifications
293T
BC008634
pCMV-ATPBD1B full-length
100 μL
Human
ATPBD1B 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 34.5kDa
Transient overexpression cell lysate was tested with anti-ATPBD1B antibody (Mfr. No. H00054707-B03) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot