Learn More
Abnova™ ATPBD1C 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00051184T01
Catalog No. 89-936-133
Add to cart
Description
Description
Specifications
Specifications
293T
NM_016301
pCMV-ATPBD1C full-length
100 μL
Human
ATPBD1C 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 31.35kDa
Transient overexpression cell lysate was tested with anti-ATPBD1C antibody (Mfr. No. H00051184-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot