Learn More
ATPG Antibody (OTI2H6), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270233H
Catalog No. NBP270233H
Add to Cart
Description
Description
ATPG Monoclonal specifically detects ATPG in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
ATPG
Monoclonal
HRP
PBS with No Preservative
ATP5F1C
Full length recombinant protein of human ATPG (NP_001001973) produced in E.coli.
0.1 mL
509.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI2H6
Western Blot, Immunohistochemistry
ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit
Mouse
Affinity Purified
Primary
Human, Monkey
IgG1
Product Suggestions
Product Suggestions