Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPG Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI2H6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270233AF405

Catalog No. N270233A405


Add to cart

Description

Description

ATPG Monoclonal antibody specifically detects ATPG in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ATPG
OTI2H6
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ATP5C1
Full length recombinant protein of human ATPG (NP_001001973) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit
Mouse
IgG1
0.1 mL
Primary
509.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit