Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATRIP Mouse anti-Canine, Human, Clone: OTI5E7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA504641
Description
ATRIP is an 86 kDa protein that is phosphorylated by ATR, regulates ATR expression, and is an essential component of the DNA damage checkpoint pathway. It localizes to the intranuclear foci after DNA damage or inhibition of replication.Specifications
ATRIP | |
Monoclonal | |
1.13 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WXE1 | |
ATRIP | |
Human recombit protein fragment corresponding to amino acids 42-309 of human ATRIP produced in E.coli. | |
100 μL | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI5E7 | |
Unconjugated | |
ATRIP | |
ATR interacting protein | |
Mouse | |
Affinity Chromatography | |
RUO | |
608504, 84126 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction