Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATRIP Mouse anti-Human, Clone: OTI9A4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504642

Catalog No. 50-167-5290


Add to cart

Description

Description

ATRIP is an 86 kDa protein that is phosphorylated by ATR, regulates ATR expression, and is an essential component of the DNA damage checkpoint pathway. It localizes to the intranuclear foci after DNA damage or inhibition of replication.
Specifications

Specifications

ATRIP
Monoclonal
1.05 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8WXE1
ATR interacting protein
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84126
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI9A4
Unconjugated
ATRIP
Liquid
ATRIP
Human recombit protein fragment corresponding to amino acids 42-309 of human ATRIP produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit