Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atropine Sulfate, Monohydrate, CP, Spectrum™

Click to view available options
Quantity:
1 kg
Packaging:
Amber Glass Bottle
Packaging Amber Glass Bottle
Quantity 1 kg
CAS 5908-99-6
CAS Max % 1
Grade Chemically Pure

CAUTION: For use by qualified personnel for research and development or other authorized use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.