Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATRX Antibody (39f), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250111AF488
Description
ATRX Monoclonal specifically detects ATRX in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ATRX | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ATRX | |
Recombinant Protein | |
0.1 mL | |
Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
546 | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
39f | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
alpha thalassemia/mental retardation syndrome X-linked, alpha thalassemia/mental retardation syndrome X-linked (RAD54 (S. cerevisiae)homolog), alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S.cerevisiae), ATP-dependent helicase ATRX, ATR2, DNA dependent ATPase and helicase, EC 3.6.1, EC 3.6.4.12, helicase 2, X-linked, Juberg-Marsidi syndrome, MGC2094, MRXHF1, RAD54 homolog, RAD54L, SFM1, SHS, transcriptional regulator ATRX, XH2RAD54, X-linked helicase II, X-linked nuclear protein, XNPZNF-HX, Zinc finger helicase, Znf-HX | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Binds N terminus of human ATRX. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction