Learn More
AFFINISEP™ AttractSPE™ Disks BIO for Molecular Biology
Used for sample cleanup of compounds from complex matrices
$521.88
Specifications
Adsorbent Type Polymeric
Base Material PTFE
Cartridge Material PTFE
Diameter 47 mm
Particle Size 10 - 20 µm
View More Specs
Products 7
Catalog Number Mfr. No. Sorbent Price Quantity & Availability  
Catalog Number Mfr. No. Sorbent Price Quantity & Availability  
BIOC1810047 View Documents AFFINISEP™
SPEDISKSBIOC18100.47.
C18 Pack of 20 for $521.88
Add to cart
BIOC4300472 View Documents AFFINISEP™
SPEDISKSBIOC4300.47.2
C4 Pack of 20 for $521.88
Add to cart
BIOC8100472 View Documents AFFINISEP™
SPEDISKSBIOC8100.47.2
C8 Bio Stage tips Protein Purification Pack of 20 for $521.88
Add to cart
BIODVB4720 View Documents AFFINISEP™
SPEDISKSBIODVB47.20
DVB Bio Stage tips Protein Purification Pack of 20 for $521.88
Add to cart
BIORPS4720 View Documents AFFINISEP™
SPEDISKSBIORPS47.20
SDB-RPS Bio Stage tips Protein Purification Pack of 20 for $521.88
Add to cart
BIOSAX4720 View Documents AFFINISEP™
SPEDISKSBIOSAX47.20
SAX Bio Stage tips Protein Purification Pack of 20 for $521.88
Add to cart
BIOSCX4720 View Documents AFFINISEP™
SPEDISKSBIOSCX47.20
SCX Bio Stage tips Protein Purification Pack of 20 for $521.88
Add to cart
Description
Description
  • AttractSPE™ Disks are based upon polymeric sorbents dedicated to sample cleanup for the extraction of compounds from complex mattrices
  • AttractSPE™ Disks are thin and uniform membranes designed for extraction, purification, and concentration of analytes from large volume liquid and air samples
  • AttractSPE™ Disks have smaller particle size of the sorbrnt material that will allow for much higher flow rates during sample loading
  • This feature makes them well suited for environmental applications
  • AttractSPE™ Disks BioMol are specifically manufactured for biological applications, such as microextractions used in staged tips, 96 well plates and mini-SPE
Specifications
Specifications
Polymeric
PTFE
10 - 20 µm
Certificate of Quality and Instruction Sheet
PTFE
47 mm
HPLC or GC
20/pk