Learn More
Invitrogen™ Attune™ NxT Focusing Fluid Filter
This 0.2 micron filter prevents contaminants from getting into the focusing fluid reservior of the Attune™ NxT Acoustic Focusing Cytometer.
Supplier: Invitrogen™ 100022587
Description
This 0.2 micron filter prevents contaminants from getting into the focusing fluid reservior of the Attune™ NxT Acoustic Focusing Cytometer. Filters are recommended for replacement once every three months, unless performing small particle detection when once-a-month replacement is recommended.
Specifications
Attune™ | |
Store at room temperature. | |
Attune Acoustic Focusing Cytometer |
Focusing Fluid Filter | |
1 Filter |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.