Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atxn1, Mouse anti-Mouse, ATTO655, Clone: S76-8, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18593

Catalog No. 89-338-518


Add to cart

Description

Description

Specifications

Specifications

Atxn1
S76-8
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user.
ataxin 1
Ataxin-1|Atx1|C85907|Sca1
Mouse
IgG2b
RUO
Antibody
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
ATTO655
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Atxn1
A synthetic peptide corresponding to amino acids 164-197 of mouse Atxn1.
100 μg
Store at -20°C.
Primary
20238
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit