Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATXN2L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550916

Catalog No. PA550916


Add to cart

Description

Description

ATXN2L Polyclonal Antibody detects endogenous levels of ATXN2L protein.

ATXN2L encodes an ataxin type 2 related protein of unknown function. This protein is a member of the spinocerebellar ataxia (SCAs) family, which is associated with a complex group of neurodegenerative disorders.
Specifications

Specifications

ATXN2L
Polyclonal
Unconjugated
ATXN2L
Liquid
ATXN2L
Synthesized peptide derived from the Internal region of human Ataxin-2L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q8WWM7
A2D, A2LG, A2LP, A2RP
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11273
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit