Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATXN2L Rabbit anti-Human, Mouse, Proteintech
Rabbit PolyClonal Antibody
Supplier: Proteintech Group Inc 248221AP150UL
Description
This gene encodes an ataxin type 2 related protein of unknown function. This protein is a member of the spinocerebellar ataxia (SCAs) family, which is associated with a complex group of neurodegenerative disorders. Several alternatively spliced transcripts encoding different isoforms have been found for this gene.Specifications
ATXN2L | |
Polyclonal | |
Unconjugated | |
ATXN2L | |
A2D, A2LG, A2LP, A2RP, Ataxin 2 domain protein, ataxin 2 like, Ataxin 2 like protein, Ataxin 2 related protein, ATXN2L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11273, 233871 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7TQH0, Q8WWM7 | |
Atxn2l | |
ATXN2L Fusion Protein Ag20581 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction