Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATXN3L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555649
Description
Immunogen sequence: LELSRQETNR EDEHLRSTIE LSMQGSSGNT SQDLPKTSCV TPASEQPKKI KEDYFEKHQQ EQKQQQQQSD LPGHSSYLHE RPTTSSRAIE SDLSDDISEG TVQAAVDTI Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 44%.
ATXN3L also named as ataxin 3 like or MJDL is a 355 amino acid protein, which contains one Josephin domain and two UIM repeats. ATXN3L localizes in nucleus and may cleave poly ubiquitin chain.
Specifications
ATXN3L | |
Polyclonal | |
Unconjugated | |
ATXN3L | |
ataxin 3 like; ataxin 3-like; Ataxin-3-like protein; ATX3L; ATXN3L; Machado-Joseph disease protein 1-like; MJDL; putative ataxin-3-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92552 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H3M9 | |
ATXN3L | |
Recombinant protein corresponding to Human ATXN3L. Recombinant protein control fragment (Product #RP-92000). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction