Learn More
Aviva Systems Biology Corp ATXN7L2 Antibody - N-terminal region (ARP53155_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53155_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATXN7L2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATXN7L2. Alternate names: FLJ00381, MGC46534ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60947