Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATXN7L2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ATXN7L2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170416
|
Novus Biologicals
NBP170416 |
100 μL |
Each for $436.00
|
|
NBP17041620
|
Novus Biologicals
NBP17041620UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
ATXN7L2 Polyclonal specifically detects ATXN7L2 in Human samples. It is validated for Western Blot.Specifications
ATXN7L2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
127002 | |
Synthetic peptides corresponding to ATXN7L2(ataxin 7-like 2) The peptide sequence was selected from the N terminal of ATXN7L2. Peptide sequence KTSSREKGQGSRSRGHQPPEKTQKDNLCQPGGLTKDSPGKPPMAPPSKEP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
ataxin 7-like 2, FLJ00381, MGC46534 | |
ATXN7L2 | |
IgG | |
Affinity Purified | |
77 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title