Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AU1 Tag Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA126547

Catalog No. PIPA126547


Add to Cart

Description

Description

PA1-26547 detects AU1 tag.

AU1 Tag Polyclonal antibody specifically detects AU1 Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AU1 Tag
Polyclonal
Unconjugated
AU1 Epitope Tag
Purified, full length, native AU1: DTYRYI conjugated to KLH.
100 μg
Primary
Antibody
IgG
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Goat
Antigen affinity chromatography
RUO
4° C
Liquid
Tag
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit