Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AU1EpitopeTag Mouse anti-Human, Clone: F7-C2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267077
Description
AU1EpitopeTag Monoclonal antibody specifically detects AU1EpitopeTag in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
AU1EpitopeTag | |
F7-C2 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40% Glycerol with 0.05% Sodium Azide | |
AU1 epitope tag, DTYRYI epitope tag, DTYRYI tag | |
Peptide | |
Affinity purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal |
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
2 mg/mL | |
Western Blot 1:2000, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
Affinity Purified | |
Mouse | |
IgG2a | |
100μL | |
Primary | |
Human, Non-species specific |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title