Learn More
Audit™ MicroControls™ MicroFD™ Tumor Markers Control
Bi-level serum-based control
Manufacturer:  Audit™ MicroControls™ K070M6
Catalog No. 22-157-141
Add to cart
This item is not returnable. View return policy
Description
Description
  • Simulate human patient samples for monitoring precision of laboratory testing procedures for AFP, CEA, CA125, CA 15-3, Free and Total PSA
  • Freeze-dried
  • Size: 6 x 2mL
Specifications
Specifications
2°C to 8°C
Simulate Human patient samples for monitoring precision of laboratory testing procedures
6 x 2 mL
alpha-Fetoprotein (AFP), CA 125, CA 15-3, Carcinoembryonic Antigen (CEA), PSA Free, PSA Total
2