Learn More
Mystaire™ Aura™ Main Filter Available on GSA/VA Contract for Federal Government customers only.
For Aura™ Series Ductless Fume Hoods. Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
Manufacturer:  Mystaire™ AU3804
 View more versions of this product
Catalog No. 15-338-117
Add to cart
Description
Description
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Main Filter
Primary
Gas Phase Carbon
Filter
Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood
Hydrocarbons