Learn More
Mystaire™ Aura™ Main Filter
For Aura™ Series Ductless Fume Hoods. Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
Manufacturer:  Mystaire™ AU2808
 View more versions of this product
Catalog No. 15-338-207
Add to cart
Description
Description
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
Specifications
Main Filter
Primary
Gas Phase Carbon
Filter
Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood
Ammonia and Amines