Learn More
Mystaire™ Aura™ Main Filter
GSA_VA
For Aura™ Series Ductless Fume Hoods. Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
$725.00 - $1006.00
Products 16
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Material Price Quantity & Availability  
15-338-181
View Documents Promotion Details
Mystaire™
AU3808
Ammonia and Amines Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $804.00
Add to cart
15-338-207
View Documents Promotion Details
Mystaire™
AU2808
Ammonia and Amines Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $875.00
Add to cart
15-338-122
View Documents Promotion Details
Mystaire™
AU3817
Custom Blend Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $1,006.00
Add to cart
15-338-115
View Documents Promotion Details
Mystaire™
AU2817
Custom Blend Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $943.00
Add to cart
15-338-118
View Documents Promotion Details
Mystaire™
AU3807
Formaldehyde Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $926.00
Add to cart
15-338-176
View Documents Promotion Details
Mystaire™
AU2807
Formaldehyde Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $866.50
Add to cart
15-338-113
View Documents Promotion Details
Mystaire™
AU2820
Particulate Air Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood HEPA
Each for $831.00
Add to cart
15-338-117
View Documents Promotion Details
Mystaire™
AU3804
Hydrocarbons Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $970.50
Add to cart
15-338-110
GSA_VA
View Documents Promotion Details
Mystaire™
AU2804
Hydrocarbons Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $785.50
Add to cart
15-338-119
View Documents Promotion Details
Mystaire™
AU3814
Mineral Acids Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $960.50
Add to cart
15-338-112
View Documents Promotion Details
Mystaire™
AU2814
Mineral Acids Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $759.50
Add to cart
15-338-121
View Documents Promotion Details
Mystaire™
AU3816
Multi-Purpose Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $945.00
Add to cart
15-338-114
View Documents Promotion Details
Mystaire™
AU2816
Multi-Purpose Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $941.00
Add to cart
15-338-120
View Documents Promotion Details
Mystaire™
AU3820
HEPA Particulates Application Aura™ 30/54 Ductless Fume Hood HEPA
Each for $833.00
Add to cart
15-338-190
View Documents Promotion Details
Mystaire™
AU3809
Sulfur Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $951.50
Add to cart
15-338-186
View Documents Promotion Details
Mystaire™
AU2809
Sulfur Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $725.00
Add to cart
Description
Description
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)