Learn More
Mystaire™ Aura™ Particulate Prefilters
GSA_VASDP
For Aura™ Series Ductless Fume Hoods. An electrostatically charged fiber filter which effectively removes submicron particulates. Designed to protect the adsorption filter from contaminants such as smoke, acid mists, and other submicron particulates.
$472.00 - $480.00
Specifications
Material Electrostatically Charged Fiber
Includes 6 Filters
Product Line Prefilter
Filter Type Prefilter
For Use With (Application) Submicron Particulates
View More Specs
Products 2
Catalog Number Mfr. No. Material Includes Product Type Product Line Filter Type For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Material Includes Product Type Product Line Filter Type For Use With (Application) Price Quantity & Availability  
15-338-116
SDP
View Documents
Mystaire™
FE2013
Electrostatically Charged Fiber 6 Filters Replacement Filter Prefilter Prefilter Submicron Particulates
Pack of 6 for $472.00
Add to cart
15-338-123
GSA_VASDP
View Documents
Mystaire™
FE3013
Electrostatically Charged Fiber 6 Filters Replacement Filter Prefilter Prefilter Submicron Particulates
Pack of 6 for $480.00
Add to cart
Description
Description
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
Specifications
Electrostatically Charged Fiber
Prefilter
Submicron Particulates
6 Filters
Prefilter