Learn More
Mystaire™ Aura™ Safety Filter
GSA_VASDP
For Aura™ Series Ductless Fume Hoods. Designed to incorporate both gas phase and mechanical phase filtration into one safety filter.
$485.00 - $855.00
Specifications
Product Type Filter
View More Specs
Products 19
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Material Price Quantity & Availability  
15-338-208
SDP
View Documents
Mystaire™
AU5208
Ammonia and Amines Aura™ 250 Ductless Fume Hood Gas Phase Carbon
Each for $485.00
Add to cart
15-338-180
SDP
View Documents
Mystaire™
AU5508
Ammonia and Amines Aura™ 42 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $503.00
Add to cart
15-338-182
SDP
View Documents
Mystaire™
AU5708
Ammonia and Amines Aura™ 42 Ductless Fume Hood Gas Phase Carbon
Each for $485.00
Add to cart
15-338-127
SDP
View Documents
Mystaire™
AU5217
Custom Blend Aura™ 250 Ductless Fume Hood Gas Phase Carbon
Each for $568.00
Add to cart
15-338-137
SDP
View Documents
Mystaire™
AU5717
Custom Blend Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $855.00
Add to cart
15-338-132
SDP
View Documents
Mystaire™
AU5517
Custom Blend Aura™ 42 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $576.00
Add to cart
15-338-177
SDP
View Documents
Mystaire™
AU5207
Formaldehyde Aura™ 250 Ductless Fume Hood Gas Phase Carbon
Each for $520.00
Add to cart
15-338-135
SDP
View Documents
Mystaire™
AU5707
Formaldehyde Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $557.00
Add to cart
15-338-130
SDP
View Documents
Mystaire™
AU5507
Formaldehyde Aura™ 42 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $488.00
Add to cart
15-338-124
SDP
View Documents
Mystaire™
AU5204
Hydrocarbons Aura™ 250 Ductless Fume Hood Gas Phase Carbon
Each for $528.00
Add to cart
15-338-134
SDP
missing translation for 'viewDocuments'
Mystaire™
AU5704
Hydrocarbons Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $554.00
Add to cart
15-338-129
GSA_VASDP
missing translation for 'viewDocuments'
Mystaire™
AU5504
Hydrocarbons Aura™ 42 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $545.00
Add to cart
15-338-156
SDP
missing translation for 'viewDocuments'
Mystaire™
AU5214
Mineral Acids Aura™ 250 Ductless Fume Hood Gas Phase Carbon
Each for $531.00
Add to cart
15-338-136
SDP
missing translation for 'viewDocuments'
Mystaire™
AU5714
Mineral Acids Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $504.00
Add to cart
15-338-131
SDP
missing translation for 'viewDocuments'
Mystaire™
AU5514
Mineral Acids Aura™ 42 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $488.00
Add to cart
15-338-170
SDP
missing translation for 'viewDocuments'
Mystaire™
AU5720
Particulate Air Aura™ 54 and Aura™ 750 Ductless Fume Hood HEPA
Each for $828.00
Add to cart
15-338-187
SDP
missing translation for 'viewDocuments'
Mystaire™
AU5209
Sulfur Aura™ 250 Ductless Fume Hood Gas Phase Carbon
Each for $697.00
Add to cart
15-338-191
SDP
missing translation for 'viewDocuments'
Mystaire™
AU5709
Sulfur Aura™ 30, Aura™ 54 and Aura™ 750 Ductless Fume Hood Gas Phase Carbon
Each for $491.00
Add to cart
15-338-189
SDP
missing translation for 'viewDocuments'
Mystaire™
AU5509
Sulfur Aura™ 42 and Aura™ 550 Ductless Fume Hood Gas Phase Carbon
Each for $525.00
Add to cart
Description
Description
  • Dual purpose safety filter is critical to ensuring operator and laboratory safety as it provides adequate reserve gas phase capacity as the primary filter(s) reach saturation.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected.
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR).
Specifications
Specifications
Filter