Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Auramine O (C.I. 41000), For Microscopy, MilliporeSigma™

Click to view available options
Quantity:
50g
2465-27-2
C17H22ClN3
303.834
KSCQDDRPFHTIRL-UHFFFAOYSA-N
Pyoktamin yellow,4, 4'-Bis-(dimethylamino)-benzophenonimide hydrochloride
17170
CHEBI:51876
4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl
2465-27-2
C17H22ClN3
303.834
KSCQDDRPFHTIRL-UHFFFAOYSA-N
Pyoktamin yellow,4, 4'-Bis-(dimethylamino)-benzophenonimide hydrochloride
17170
CHEBI:51876
4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl
Appearance Yellow solid
UN Number UN 3077
Formula Weight 303.84
Physical Form Solid
Packaging Glass bottle
Quantity 50g
Chemical Name or Material Auramine O (C.I. 41000)

H302: Harmful if swallowed.
H319: Causes serious eye irritation.
H351: Suspected of causing cancer.
H411: Toxic to aquatic life with long lasting effects.