Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Auramine O (Certified Biological Stain), Fisher Chemical

Catalog Number a96825
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
2465-27-2
C17H22ClN3
303.834
MFCD00012484
KSCQDDRPFHTIRL-UHFFFAOYSA-N
Basic Yellow 2
17170
CHEBI:51876
4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl
2465-27-2
C17H22ClN3
303.834
MFCD00012484
KSCQDDRPFHTIRL-UHFFFAOYSA-N
Basic Yellow 2
17170
CHEBI:51876
4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl
For use in fluorescence microscopy.
TRUSTED_SUSTAINABILITY
Melting Point 267.2°C
Bio. Stain Comm. Certification Pass Test
Dye Content Report %
Formula Weight 303.84
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 41000
Color Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Auramine O
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit