Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Auranofin, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
34031-32-8
C20H34AuO9PS
678.483
AUJRCFUBUPVWSZ-UHFFFAOYSA-M
4-2-aminoethyl benzene sulfonyl fluoride hydrochloride
24199313
gold(1+);3,4,5-triacetyloxy-6-(acetyloxymethyl)oxane-2-thiolate;triethylphosphane
CCP(CC)CC.CC(=O)OCC1C(C(C(C(O1)[S-])OC(=O)C)OC(=O)C)OC(=O)C.[Au+]
This item is not returnable. View return policy
34031-32-8
C20H34AuO9PS
678.483
AUJRCFUBUPVWSZ-UHFFFAOYSA-M
4-2-aminoethyl benzene sulfonyl fluoride hydrochloride
24199313
gold(1+);3,4,5-triacetyloxy-6-(acetyloxymethyl)oxane-2-thiolate;triethylphosphane
CCP(CC)CC.CC(=O)OCC1C(C(C(C(O1)[S-])OC(=O)C)OC(=O)C)OC(=O)C.[Au+]

Thioredoxin reductase inhibitor; induces MPT

Inhibitor of thioredoxin reductase (TrxR) (IC50 = 20 nM; Ki = 4 nM for the NADPH-reduced form of human cytosolic TrxR). Thought to induce the mitochondrial permeability transition via inhibition of mitochondrial TrxR. Also exhibits anti-inflammatory and immunosuppressive activities; inhibits 5-lipoxygenase at high concentrations and stimulates LTA hydrolase at low concentrations.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 678.48
Percent Purity >98%
Chemical Name or Material Auranofin

Recommended Storage : Store at Room Temperature