Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora A Monoclonal Antibody (35C1), Active Motif™
Mouse Monoclonal Antibody
$147.50 - $565.00
Specifications
Antigen | Aurora A |
---|---|
Clone | 35C1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3643
|
Active Motif
61710 |
10 μg |
Each for $147.50
|
|
|||||
50-199-3642
|
Active Motif
61709 |
100 μg |
Each for $565.00
|
|
|||||
Description
Aurora A is a 46 kDa protein that plays a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal. Aurora A is viewed as a potential target for anticancer drug treatment.Tissue specificity: Highly expressed in testis and weakly in skeletal muscle, thymus and spleen. Also highly expressed in colon, ovarian, prostate, neuroblastoma, breast and cervical cancer cell lines. Multiple transcript variants encoding the same protein have been found for this gene. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal.Specifications
Aurora A | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
Aik; Airk; AIRK1; Ark1; ARK-1; AU019385; AURA; AURAK; AURKA; Aurora 2; aurora A; Aurora A Kinase; Aurora family kinase 1; Aurora kinase A; aurora/IP; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; AURORA2; Aurora-A; aurora-related kinase 1; AW539821; Ayk1; BOS_13320; Breast tumor-amplified kinase; BTAK; CR956640.2; EC 2.7.11.1; hARK1; IAK; Iak1; Ipl1- and aurora-related kinase 1; IPL1-related kinase; MGC34538; OTTHUMP00000031341; OTTHUMP00000166072; PPP1R47; protein phosphatase 1, regulatory subunit 47; ratAurA; RP5-1167H4.6; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase Ayk1; Stk15; Stk6; STK7 | |
AURKA | |
IgG2b | |
Protein A | |
Antibody |
35C1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O14965 | |
6790 | |
This antibody was raised against full-length recombinant human Aurora A protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
AURKA |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title