Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora B Mouse anti-Human, Clone: AURKB/1521, Novus Biologicals™
Mouse Monoclonal Antibody
$240.50 - $482.00
Specifications
Antigen | Aurora B |
---|---|
Clone | AURKB/1521 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25326720
|
Novus Biologicals
NBP25326720UG |
20 μg |
Each for $240.50
|
|
|||||
NB253267100
|
Novus Biologicals
NBP253267100UG |
100 μg |
Each for $482.00
|
|
|||||
NB253267200
|
Novus Biologicals
NBP253267200UG |
200 μg | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
Aurora B Monoclonal specifically detects Aurora B in Human samples. It is validated for Western Blot.Specifications
Aurora B | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
9212 | |
Recombinant human Aurora B protein fragment (aa89-251) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
AURKB/1521 | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
AURKB | |
IgG2b | |
Protein A or G purified | |
39 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title