Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora B Mouse anti-Human, PE-Cyanine7, Clone: 2B1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP174027PECY7
Description
Aurora B Monoclonal antibody specifically detects Aurora B in Human samples. It is validated for Flow Cytometry.Specifications
Aurora B | |
2B1 | |
Flow Cytometry | |
Conjugated, Purified | |
AURKB | |
Recombinant human Aurora kinase B (1-344aa) purified from E. coli | |
Protein G purified | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
Primary | |
9212.0 | |
Human |
Flow Cytometry | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Anti-human Aurora kinase B mAb, clone 2B1, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Aurora kinase B protein. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title