Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora B Rabbit anti-Human, Clone: AURKB/3121R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Aurora B
Clone AURKB/3121R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122582
View Documents
Novus Biologicals
NBP30755420UG
20 μg
Each for $159.82
Add to cart
 
NB122581
View Documents
Novus Biologicals
NBP30755410UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Aurora B Monoclonal antibody specifically detects Aurora B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Aurora B
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5
Recombinant fragment (around aa 89-251) of human Aurora B protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
AURKB/3121R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers
10 mM PBS with 0.05% BSA
9212
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit