Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora C Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI2B7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271303AF700

Catalog No. N271303A700


Add to cart

Description

Description

Aurora C Monoclonal antibody specifically detects Aurora C in Human samples. It is validated for Western Blot.
Specifications

Specifications

Aurora C
OTI2B7
Western Blot
Affinity Purified, Conjugated
AURKC
Full-length protein expressed in 293T cell transfected with human Aurora C expression vector.
Immunogen affinity purified
Cell Cycle and Replication
Primary
6795.0
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
AIE2AIK3, ARK3Serine/threonine-protein kinase aurora-C, AurC, aurora kinase CAurora/IPL1/Eg2 protein 2, Aurora/IPL1-related kinase 3, aurora-C, Aurora-related kinase 3, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 13 (aurora/IPL1-like), serine/threonine-protein kinase 13, STK13ARK-3
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit