Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific Autoclavable Bags

Clear, imprinted bags with blue biohazard labeling specifically designed for safe disposal

$122.15 - $251.32

Specifications

Autoclavable Autoclavable
Color Clear
Description Clear Autoclavable Bags
Disposable Yes
For Use With (Application) Hazardous Waste Disposal
View More Specs

Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
03-448-267
View Documents
Heathrow Scientific
HS1002A
310 x 660 mm
Case of 200 for $122.15
Add to cart
 
03-448-268
View Documents
Heathrow Scientific
HS1002B
415 x 600 mm
Case of 200 for $132.17
Add to cart
 
03-448-269
View Documents
Heathrow Scientific
HS1002C
610 x 810 mm
Case of 200 for $251.32
Add to cart
 
Description

Description

  • Constructed of 2.0 mm thick polypropylene
  • Can be used for repeated autoclaving
  • Strong side seams securely hold items
  • Autoclavable at 250°F (121°C)
  • Front write-on panel
  • 4 inner packs of 50 bags
  • Disposable
Specifications

Specifications

Autoclavable
Clear Autoclavable Bags
Hazardous Waste Disposal
Polypropylene
50/Pk.
Non-sterile
Autoclavable at 250°F
Clear
Yes
Autoclave
200/Cs
Box
Room Temperature
0.08 in.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit