Learn More
Andwin ScientificSupplier Diversity Partner AUTOCLAVE BAG 38X48 3MIL 50/CS  DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 14220088
Bag; Biohazard; Double Thick Autoclavable; 96.5 x 122cm; Heavy Duty 3mil; Polypropylene; Red; Combines extra puncture resistance and high impact strength; Autoclavable up to 135 deg.C; Printed biohazard warning labeling
Catalog No. NC9361052